D(-)-Arabinose

Lieferant: Molekula

MOLEM18395715EA 0 EUR
MOLEM18395715
D(-)-Arabinose
D(-)-Arabinose
Formel: C₅H₁₀O₅
Molecular Weight: 150,13 g/mol
Schmelzpunkt: 160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00135608
CAS-Nummer: 10323-20-3
EINECS: 233-708-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR