Streptomycinsulfat, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ200
SIALS1277-5GEA
35.5
EUR
SIALS1277-5G
SIALS1277-50G
Streptomycinsulfat, Sigma-Aldrich®
Streptomycin sulphate
Formel:
C₂₁H₃₉N₇O₁₂·1.5H₂SO₄ Molecular Weight: 1457,4 g/mol Schmelzpunkt: 250 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00037023 CAS-Nummer: 3810-74-0 EINECS: 223-286-0 UN: 2811 ADR: 6.1,III Merck Index: 12,08983 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...