3-Mercapto-1,2-propandiol
Lieferant: Serva
|
Achtung
|
Synonyme:
(±)-1-Thioglycerin
, (±)-α-Monothioglycerin
, (±)-3-Mercapto-1,2-dihydroxypropan
SERA28637.02EA
0
EUR
SERA28637.02
SERA28637.01
3-Mercapto-1,2-propandiol
3-Mercapto-1,2-propandiol
Formel:
C₃H₈O₂S Molecular Weight: 108,16 g/mol Siedepunkt: 118 °C (1013 hPa) Dichte: 1,247 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004879 CAS-Nummer: 96-27-5 EINECS: 202-495-0 Merck Index: 12,09471 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...