Dithiothreitol (DTT, Cleland's Reagenz) 1 mol/l in wässriger Lösung

Lieferant: UPSTATE TECHNOLOGY

Synonyme: DTT(R*,R*)-1,4-Dimercapto-2,3-butandiol

UPST20-265EA 0 EUR
UPST20-265
Dithiothreitol (DTT, Cleland's Reagenz) 1 mol/l in wässriger Lösung
Dithiothreitol (DTT, Cleland's Reagenz)
Formel: C₄H₁₀O₂S₂
Molecular Weight: 154,25 g/mol
MDL: MFCD00004877
CAS-Nummer: 3483-12-3
EINECS: 222-468-7
https://de.vwr.com/stibo/web/std.lang.all/98/19/2299819.jpg

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR