Ethidiumbromid 10 mg/ml in wässriger Lösung
Lieferant: Uptima
|
Gefahr
|
Synonyme:
3,8-Diamino-5-ethyl-6-phenylphenanthridiniumbromid
UPTIUP89244BEA
0
EUR
UPTIUP89244B
Ethidiumbromid 10 mg/ml in wässriger Lösung
Ethidiumbromid
Formel:
C₂₁H₂₀BrN₃ Molecular Weight: 394,31 g/mol |
MDL:
MFCD00011724 CAS-Nummer: 1239-45-8 EINECS: 214-984-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...