Talk
Lieferant: CAESAR & LORENTZ GMBH
|
Achtung
|
Synonyme:
Talkum
, Magnesiumsilikat Monohydrat
, Magnesiumsilikathydrat
CSLO2660-1KGEA
24.7
EUR
CSLO2660-1KG
Talk
Talk
Formel:
Mg₃O₁₁Si₄·1H₂O Molecular Weight: 379,27 g/mol Schmelzpunkt: >1200 °C Dichte: 2,58…2.83 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00792903 CAS-Nummer: 14807-96-6 EINECS: 238-877-9 Merck Index: 13,09127 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...