Mangan(II)chlorid Monohydrat

Lieferant: Honeywell Chemicals
Achtung

HONE13297.9030EA 0 EUR
HONE13297.9030
Mangan(II)chlorid Monohydrat
Mangan(II)chlorid Monohydrat
Formel: Cl₂Mn·1H₂O
Molecular Weight: 143,86 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 64333-01-3
EINECS: 231-869-6
UN: 3077
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR