Cobalt(II)nitrat Hexahydrat, EMSURE® ACS, Reag. Ph. Eur. zur Analyse (max. 0,001% Ni), Supelco®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Cobaltdinitrat Hexahydrat
, Cobaltnitrat Hexahydrat
M-Clarity™ Qualitätsstufe = MQ300
1.02554.0250EA
519
EUR
1.02554.0250
1.02554.9999
1.02554.0050
Cobalt(II)nitrat Hexahydrat, EMSURE® ACS, Reag. Ph. Eur. zur Analyse (max. 0,001% Ni), Supelco®
Cobalt(II)nitrat Hexahydrat
Formel:
Co(NO₃)₂·6H₂O Molecular Weight: 291,03 g/mol Schmelzpunkt: 57 °C Dichte: 1,88 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149647 CAS-Nummer: 10026-22-9 EINECS: 233-402-1 UN: 3077 ADR: 9,III Merck Index: 13,02469 |
Spezifikation Testergebnisse
Solubility | 2170 g/l (100 °C) |
Melting Point | 57 °C |
Molar Mass | 291.04 g/mol |
Bulk Density | 800 kg/m3 |
Density | 1.87 g/cm3 (20 °C) |
pH | 4.0 (100 g/l, H2O, 20 °C) |
Hazard Class | 9 |
Hazard Class Description | Miscellaneous Hazardous Material |
Hazard Class Description | Miscellaneous Hazardous Material |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...