D(-)-Glutamin

Lieferant: Molekula

Synonyme: D(-)-2-Amino-1,5-pentandisäure-5-amidD(-)-Glutaminsäure-5-amid(R)-(-)-Glutamin(R)-(-)-2-Amino-1,5-pentandisäure-5-amid(R)-(-)-Glutaminsäure-5-amidH-D-Gln-OH

MOLEM58264131EA 0 EUR
MOLEM58264131
D(-)-Glutamin
D(-)-Glutamin
Formel: C₅H₁₀N₂O₃
Molecular Weight: 146,15 g/mol
MDL: MFCD00065607
CAS-Nummer: 5959-95-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR