(S)-(+)-1-Methoxy-2-propanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-(+)-Propylenglycolmonomethylether

SIAL88808-1ML-FEA 132 EUR
SIAL88808-1ML-F
(S)-(+)-1-Methoxy-2-propanol, Sigma-Aldrich®
(S)-(+)-1-Methoxy-2-propanol
Formel: C₄H₁₀O₂
Molecular Weight: 90,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01632588
CAS-Nummer: 26550-55-0
UN: 3092
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR