D(-)-allo-Isoleucin

Lieferant: Molekula

Synonyme: (2R,3S)-(-)-α-Amino-β-methylvaleriansäure(2R,3S)-(-)-2-Amino-3-methylpentansäureH-D-allo-Ile-OH(2R,3S)-(-)-allo-Isoleucin

MOLEM23313421EA 0 EUR
MOLEM23313421
D(-)-allo-Isoleucin
D(-)-allo-Isoleucin
Formel: C₆H₁₃NO₂
Molecular Weight: 131,17 g/mol
MDL: MFCD00066445
CAS-Nummer: 1509-35-9
EINECS: 216-143-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR