L(+)-Isoleucin, Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyme:
(2S,3S)-(+)-2-Amino-3-methylpentansäure
, (2S,3S)-(+)-α-Amino-β-methylvaleriansäure
, (2S,3S)-(+)-Isoleucin
4160-100EA
248
EUR
4160-100
L(+)-Isoleucin, Millipore®
L(+)-Isoleucin
Formel:
H₃CCH₂CH(CH₃)CH(NH₂)COOH Molecular Weight: 131,17 g/mol Schmelzpunkt: 279…280 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064222 CAS-Nummer: 73-32-5 EINECS: 200-798-2 Merck Index: 14,05179 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...