L(+)-Isoleucin

Lieferant: Molekula

Synonyme: (2S,3S)-(+)-2-Amino-3-methylpentansäure(2S,3S)-(+)-α-Amino-β-methylvaleriansäure(2S,3S)-(+)-Isoleucin

MOLE15495360-100GEA 68.7 EUR
MOLE15495360-100G
L(+)-Isoleucin
L(+)-Isoleucin
Formel: H₃CCH₂CH(CH₃)CH(NH₂)COOH
Molecular Weight: 131,17 g/mol
Schmelzpunkt: 279…280 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064222
CAS-Nummer: 73-32-5
EINECS: 200-798-2
Merck Index: 13,05198

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR