Kalium-tert-butanolat

Lieferant: Molekula
Gefahr

Synonyme: Kalium-tert-butanolatKalium-tert-butylat2-Methyl-2-propanol Kaliumsalz

MOLE12739396-500GEA 232 EUR
MOLE12739396-500G
Kalium-tert-butanolat
Kalium-tert-butanolat
Formel: C₄H₉KO
Molecular Weight: 112,21 g/mol
Schmelzpunkt: 256…258 °C
MDL: MFCD00012162
CAS-Nummer: 865-47-4
EINECS: 212-740-3
UN: 3206
ADR: 4.2,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR