Isoamylalkohol ≥99,0%, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals
Achtung

Synonyme: 3-Methyl-1-butanolGärungsamylalkoholIsopentylalkoholiso-Pentanol

20798.295EA 105 EUR
20798.295
Isoamylalkohol ≥99,0%, AnalaR NORMAPUR® analytisches Reagens
Isoamylalkohol
Formel: (CH₃)₂CHCH₂CH₂OH
Molecular Weight: 88,15 g/mol
Siedepunkt: 130 °C (1013 hPa)
Schmelzpunkt: –117 °C
Dichte: 0,812 g/cm³ (20 °C)
Flammpunkt: 42 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002934
CAS-Nummer: 123-51-3
EINECS: 204-633-5
UN: 1105
ADR: 3,III
Merck Index: 13,05214
https://de.vwr.com/stibo/web/std.lang.all/15/69/2301569.jpg

Bestellen

Spezifikation Testergebnisse

Assay (calculated as mixture of isomers) Min. 99.0 %
Acidity Max. 0.008 meq/g
Alkalinity Max. 0.0008 meq/g
Boiling point 131 - 132.5 °C
Colouration Max. 10 APHA
Density (20/4) 0.808 - 0.812
Acidity and esters (as C7H14O2) Max. 0,17 %
Evaporation residue Max. 10 ppm
Furfuraldehyde Max. 1 ppm
Pentan-1-ol Max. 0.5 %
Peroxides (as H2O2) Max. 10 ppm
Water Max. 0.2 %
Al (Aluminium) Max. 0.5 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.02 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.1 ppm
Sn (Tin) Max. 0.1 ppm
Zn (Zinc) Max. 0.1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR