(4-Aminophenyl)essigsäure
Lieferant: Molekula
MOLE12062232-25GEA
0
EUR
MOLE12062232-25G
(4-Aminophenyl)essigsäure
(4-Aminophenyl)essigsäure
Formel:
(C₆H₄(NH₂))CH₂COOH Molecular Weight: 151,16 g/mol Siedepunkt: 322 °C (1013 hPa) Schmelzpunkt: 199…202 °C |
MDL:
MFCD00007916 CAS-Nummer: 1197-55-3 EINECS: 214-828-7 Merck Index: 13,00462 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...