HEPES Natriumsalz
Lieferant: LGC Standards PROMOCHEM
Synonyme:
2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure Natriumsalz
PROONIST-2182EA
948
EUR
PROONIST-2182
HEPES Natriumsalz
HEPES Natriumsalz
Formel:
C₈H₁₇N₂NaO₄S Molecular Weight: 260,29 g/mol Schmelzpunkt: ? 105 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036463 CAS-Nummer: 75277-39-3 EINECS: 278-169-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...