Acrylamid ≥99,9%, ultrareines Bioreagenz für die Elektrophorese, gereinigt, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Acrylsäureamid2-PropenamidPropensäureamidPropenamid

4081-01EA 0 EUR
4081-01
Acrylamid ≥99,9%, ultrareines Bioreagenz für die Elektrophorese, gereinigt, J.T.Baker®
Acrylamid
Formel: C₃H₅NO
Molecular Weight: 71,08 g/mol
Siedepunkt: 232 °C (1013 hPa)
Schmelzpunkt: 84,5 °C
Dichte: 1,122 g/cm³ (20 °C)
Flammpunkt: 138 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00008032
CAS-Nummer: 79-06-1
EINECS: 201-173-7
UN: 2074
ADR: 6.1,III
https://de.vwr.com/stibo/web/std.lang.all/13/21/2301321.jpg

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use
Store between 18°C and 26°C.
Assay (CH₂:CHCONH₂) ≥ 99.9 %
pH of 10% Solution in 0.1M NaCl 6.0 - 7.0
Insoluble in H₂O ≤ 0.005 %
Insoluble in Methanol ≤ 0.005 %
Acrylic Acid (CH₂:CHCOOH) ≤ 0.001 %
Absorbance of a 1% Solution at 290 nm (1-cm path vs water)(au) ≤ 0.100
Conductivity of 35% Solution, µmho ≤ 2.5
RNase Activity Passes Test
DNase Activity (None Detected) Passes Test
Protease Activity Passes Test
Heavy Metals (as Pb) ≤ 1 ppm
Iron (Fe) ≤ 1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR