2,4-Dichlorzimtsäure

Lieferant: FLUOROCHEM LTD (CHEMPUR)

FLUO011139-50GEA 136 EUR
FLUO011139-50G
2,4-Dichlorzimtsäure
2,4-Dichlorzimtsäure
Formel: C₉H₆Cl₂O₂
Molecular Weight: 217,04 g/mol
MDL: MFCD00004373
CAS-Nummer: 20595-45-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR