5-Iodisatin

Lieferant: MENAI (PART OF CHEMPUR)

Synonyme: 5-Jodindolin-2,3-dion5-Jod-1H-indol-2,3-dion5-Iodindolin-2,3-dion5-Iod-1H-indol-2,3-dion5-Jod-2,3-indolindion5-Jod-2,3-dioxoindolin5-Iod-2,3-dioxoindolin5-Iod-2,3-indolindion

MENAE2692.5EA 0 EUR
MENAE2692.5 MENAE2692.25
5-Iodisatin
5-Iodisatin
Formel: C₈H₄INO₂
Molecular Weight: 273,03 g/mol
MDL: MFCD00016899
CAS-Nummer: 20780-76-1
EINECS: 244-035-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR