3-Bromphenylisothiocyanat
Lieferant: Thermo Scientific
Synonyme:
1-Bromo-3-isothiocyanatobenzene
S04141DA
S04141EA
S04141EE
MAYBS04141DAEA
0
EUR
MAYBS04141DA
MAYBS04141EA
MAYBS04141EE
3-Bromphenylisothiocyanat
3-Bromphenylisothiocyanat
Formel:
C₇H₄BrNS Molecular Weight: 214,09 g/mol Siedepunkt: 256 °C (1013 hPa) |
MDL:
MFCD00004803 CAS-Nummer: 2131-59-1 UN: 3335 ADR: 9, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...