Methyl-4-chlorbenzoat
Lieferant: FLUOROCHEM LTD (CHEMPUR)
Synonyme:
4-Chlorbenzoesäuremethylester
FLUO018826-25GEA
29.4
EUR
FLUO018826-25G
Methyl-4-chlorbenzoat
Methyl-4-chlorbenzoat
Formel:
C₈H₇ClO₂ Molecular Weight: 170,6 g/mol Siedepunkt: 103…106 °C (15 mmHg) Schmelzpunkt: 43 °C Dichte: 1,382 g/cm³ (20 °C) |
MDL:
MFCD00000621 CAS-Nummer: 1126-46-1 EINECS: 214-420-9 Merck Index: 13,02144 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...