2-Chlorphenylisothiocyanat

Lieferant: Thermo Scientific

MAYBSB01592.1EA 0 EUR
MAYBSB01592.1 MAYBSB01592.50
2-Chlorphenylisothiocyanat
2-Chlorphenylisothiocyanat
Formel: C₇H₄ClNS
Molecular Weight: 169,63 g/mol
Siedepunkt: 260 °C (1013 hPa)
Dichte: 1,29 g/cm³ (20 °C)
MDL: MFCD00004801
CAS-Nummer: 2740-81-0
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR