2-Chlorphenylisothiocyanat

Lieferant: FLUOROCHEM LTD (CHEMPUR)

FLUO008773-25GEA 114 EUR
FLUO008773-25G FLUO008773-5G FLUO008773-10G
2-Chlorphenylisothiocyanat
2-Chlorphenylisothiocyanat
Formel: C₇H₄ClNS
Molecular Weight: 169,63 g/mol
Siedepunkt: 260 °C (1013 hPa)
Dichte: 1,29 g/cm³ (20 °C)
MDL: MFCD00004801
CAS-Nummer: 2740-81-0
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR