2,4-Dichlorbenzolsulfonylchlorid

Lieferant: MENAI (PART OF CHEMPUR)

Synonyme: 2,4-Dichlorbenzolsulfochlorid2,4-Dichlorbenzolsulfonsäurechlorid2,4-Dichlorbenzol-1-sulfonylchlorid

MENANC1149.25EA 0 EUR
MENANC1149.25
2,4-Dichlorbenzolsulfonylchlorid
2,4-Dichlorbenzolsulfonylchlorid
Formel: C₆H₃Cl₃O₂S
Molecular Weight: 245,51 g/mol
Schmelzpunkt: 56 °C
MDL: MFCD00052712
CAS-Nummer: 16271-33-3
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR