Ethyl-3-fluorbenzoat

Lieferant: CHEMPUR

Synonyme: 3-Fluorbenzoesäureethylester

FL6107.50 FL6107.10 FL6107.250
CHMPFL6107.50EA 0 EUR
CHMPFL6107.50 CHMPFL6107.10 CHMPFL6107.250
Ethyl-3-fluorbenzoat
Ethyl-3-fluorbenzoat
Formel: C₉H₉FO₂
Molecular Weight: 168,17 g/mol
MDL: MFCD00000336
CAS-Nummer: 451-02-5
EINECS: 207-191-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR