4-Fluor-2-nitrobenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL115452-1GEA 58.2 EUR
SIAL115452-1G
4-Fluor-2-nitrobenzoesäure, Sigma-Aldrich®
4-Fluoro-2-nitrobenzoic acid
Formel: C₇H₄FNO₄
Molecular Weight: 185,11 g/mol
MDL: MFCD00024300
CAS-Nummer: 394-01-4
EINECS: 206-890-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR