Resveratrol
Lieferant: UPSTATE TECHNOLOGY
UPST19-172EA
0
EUR
UPST19-172
Resveratrol
Resveratrol
Formel:
C₁₄H₁₂O₃ Molecular Weight: 228,25 g/mol Schmelzpunkt: 260 °C Dichte: 1,36 g/cm³ (20 °C) Lagertemperatur: Gefrierschrank |
MDL:
MFCD00133799 CAS-Nummer: 501-36-0 Merck Index: 12,08326 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...