ω-Brom-3,4-dichloracetophenon

Lieferant: MENAI (PART OF CHEMPUR)

Synonyme: α-Brom-3,4-dichloracetophenon2-Brom-3',4'-dichloracetophenon3,4-Dichlorphenacylbromid2-Brom-1-(3,4-dichlorphenyl)ethan-1-on

MENAE2681.10EA 0 EUR
MENAE2681.10
ω-Brom-3,4-dichloracetophenon
ω-Brom-3,4-dichloracetophenon
Formel: C₈H₅BrCl₂O
Molecular Weight: 267,94 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00051581
CAS-Nummer: 2632-10-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR