Pentamethylbenzolsulfonylchlorid

Lieferant: FLUOROCHEM LTD (CHEMPUR)

Synonyme: PentamethylbenzolsulfochloridPentamethylbenzolsulfonsäurechloridPentamethylbenzol-1-sulfonylchlorid

FLUO018935-5GEA 0 EUR
FLUO018935-5G
Pentamethylbenzolsulfonylchlorid
Pentamethylbenzolsulfonylchlorid
Formel: C₁₁H₁₅ClO₂S
Molecular Weight: 246,76 g/mol
Schmelzpunkt: 82 °C
MDL: MFCD00014722
CAS-Nummer: 52499-94-2
EINECS: 257-968-4
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR