Pentamethylbenzolsulfonylchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: PentamethylbenzolsulfochloridPentamethylbenzolsulfonsäurechloridPentamethylbenzol-1-sulfonylchlorid

M-Clarity™ quality level = MQ100

SIAL565369-5GEA 50.5 EUR
SIAL565369-5G
Pentamethylbenzolsulfonylchlorid, Sigma-Aldrich®
Pentamethylbenzolsulfonylchlorid
Formel: C₁₁H₁₅ClO₂S
Molecular Weight: 246,76 g/mol
Schmelzpunkt: 82 °C
MDL: MFCD00014722
CAS-Nummer: 52499-94-2
EINECS: 257-968-4
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR