2,2',3,4,4',6'-Hexachlorbiphenyl (PCB Nr. 140)

Lieferant: LGC Standards PROMOCHEM

PROOCIL-PCB-140-CEA 652 EUR
PROOCIL-PCB-140-C
2,2',3,4,4',6'-Hexachlorbiphenyl (PCB Nr. 140)
2,2',3,4,4',6'-Hexachlorbiphenyl (PCB Nr. 140)
Formel: C₁₂H₄Cl₆
Molecular Weight: 360,88 g/mol
CAS-Nummer: 59291-64-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR