2,2',3,4',5',6-Hexachlorbiphenyl (PCB Nr. 149)

Lieferant: LGC Standards PROMOCHEM

PROOCIL-PCB-149-CEA 687 EUR
PROOCIL-PCB-149-C
2,2',3,4',5',6-Hexachlorbiphenyl (PCB Nr. 149)
2,2',3,4',5',6-Hexachlorbiphenyl (PCB Nr. 149)
Formel: C₁₂H₄Cl₆
Molecular Weight: 360,88 g/mol
CAS-Nummer: 38380-04-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR