3-Chlorbiphenyl (PCB Nr. 2)

Lieferant: LGC Standards PROMOCHEM

PROOCIL-PCB-2-CEA 560 EUR
PROOCIL-PCB-2-C
3-Chlorbiphenyl (PCB Nr. 2)
3-Chlorbiphenyl (PCB Nr. 2)
Formel: C₁₂H₉Cl
Molecular Weight: 188,66 g/mol
Siedepunkt: 285 °C (1013 hPa)
Schmelzpunkt: 16 °C
Dichte: 1,1579 g/cm³ (25 °C)
MDL: MFCD00013633
CAS-Nummer: 2051-61-8
EINECS: 218-126-1
UN: 2315
ADR: 9,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR