4,4'-Biphenol
Lieferant: EHRENSTORFER GMBH
Synonyme:
1-Hydroxy-4-(4-hydroxyphenyl)benzol
, Biphenyl-4,4'-diol
, 4,4'-Dihydroxybiphenyl
EHERC12635500EA
51
EUR
EHERC12635500
4,4'-Biphenol
4,4'-Biphenol
Formel:
C₁₂H₁₀O₂ Molecular Weight: 186,21 g/mol Siedepunkt: 355 °C (1013 hPa) Dichte: 1,25 g/cm³ (20 °C) |
MDL:
MFCD00002348 CAS-Nummer: 92-88-6 EINECS: 202-200-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...