6-Methylycumarin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Methyl-2-benzopyron

M-Clarity™ quality level = MQ100

SIALM36203-5GEA 33.9 EUR
SIALM36203-5G
6-Methylycumarin, Sigma-Aldrich®
6-Methylycumarin
Formel: C₁₀H₈O₂
Molecular Weight: 160,17 g/mol
Siedepunkt: 303 °C (1013 hPa)
Schmelzpunkt: 72…74 °C
MDL: MFCD00006875
CAS-Nummer: 92-48-8
EINECS: 202-158-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR