4-Ethylcyclohexanon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL129380-25GEA 112 EUR
SIAL129380-25G
4-Ethylcyclohexanon, Sigma-Aldrich®
4-Ethylcyclohexanon
Formel: C₈H₁₄O
Molecular Weight: 126,2 g/mol
Siedepunkt: 192 °C (1013 hPa)
Dichte: 0,91 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001644
CAS-Nummer: 5441-51-0
EINECS: 226-629-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR