2-Brom-9-fluorenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL300128-100MGEA 94.3 EUR
SIAL300128-100MG
2-Brom-9-fluorenon, Sigma-Aldrich®
2-Brom-9-fluorenon
Formel: C₁₃H₇BrO
Molecular Weight: 259,1 g/mol
MDL: MFCD00010789
CAS-Nummer: 3096-56-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR