Xanthydrol ≥98%
Lieferant: Thermo Scientific
Synonyme:
9-Hydroxyxanthen
, 9H-Xanthen-9-ol
, 9-Xanthenol
A13476.14EA
196
EUR
A13476.14
A13476.06
Xanthydrol ≥98%
Xanthydrol
Formel:
C₁₃H₁₀O₂ Molecular Weight: 198,22 g/mol Siedepunkt: 338 °C (1013 hPa) Schmelzpunkt: 122…126 °C |
MDL:
MFCD00005057 CAS-Nummer: 90-46-0 EINECS: 201-996-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...