Xanthydrol

Lieferant: Molekula

Synonyme: 9-Hydroxyxanthen9H-Xanthen-9-ol9-Xanthenol

MOLEM82885570EA 0 EUR
MOLEM82885570
Xanthydrol
Xanthydrol
Formel: C₁₃H₁₀O₂
Molecular Weight: 198,22 g/mol
Siedepunkt: 338 °C (1013 hPa)
Schmelzpunkt: 123 °C
MDL: MFCD00005057
CAS-Nummer: 90-46-0
EINECS: 201-996-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR