5-Ethyl-2-furaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL186996-1GEA 68.6 EUR
SIAL186996-1G
5-Ethyl-2-furaldehyd, Sigma-Aldrich®
5-Ethyl-2-furaldehyd
Formel: C₇H₈O₂
Molecular Weight: 124,14 g/mol
Siedepunkt: 88…89 °C (14 mmHg)
Dichte: 1,05 g/cm³ (20 °C)
MDL: MFCD00030605
CAS-Nummer: 23074-10-4
EINECS: 245-411-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR