Kaliumtetrafluoroborat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL278955-500GEA
79.3
EUR
SIAL278955-500G
Kaliumtetrafluoroborat, Sigma-Aldrich®
Potassium tetrafluoroborate
Formel:
BF₄K Molecular Weight: 125,9 g/mol Siedepunkt: 960,85 °C (310 mmHg) Schmelzpunkt: 96,5…98 °C Dichte: 3,348 g/cm³ (25 °C) |
MDL:
MFCD00011395 CAS-Nummer: 14075-53-7 EINECS: 237-928-2 Merck Index: 12,07859 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...