Kaliumhexacyanoferrat(II)

Lieferant: YSI LIFE SCIENCE PRODUCTS

YSIL2363EA 0 EUR
YSIL2363
Kaliumhexacyanoferrat(II)
Kaliumhexacyanoferrat(II)
Formel: C₆FeK₄N₆
Molecular Weight: 368,35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00167022
CAS-Nummer: 13943-58-3
EINECS: 237-722-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR