Ammoniumhydrogendifluorid
Lieferant: BIONET RESEARCH (CHEMPUR)
|
Gefahr
|
Synonyme:
Ammoniumbifluorid
, Ammoniumhydrogenfluorid
BIOT000390-1000EA
0
EUR
BIOT000390-1000
Ammoniumhydrogendifluorid
Ammoniumhydrogendifluorid
Formel:
NH₄HF₂ Molecular Weight: 57,04 g/mol Schmelzpunkt: 127 °C Dichte: 0,8 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012018 CAS-Nummer: 1341-49-7 EINECS: 215-676-4 UN: 1727 ADR: 8,II Merck Index: 13,00498 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...