(3-(Dimethylamino)propyl)trimethoxysilan, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
N,N-Dimethyl-3-(trimethoxysilyl)propylamin
, Trimethoxy(3-(dimethylamino)propyl)silan
, (3-Dimethylaminopropyl)trimethoxysilan
M-Clarity™ quality level = MQ100
SIAL539287-25MLEA
124
EUR
SIAL539287-25ML
(3-(Dimethylamino)propyl)trimethoxysilan, Sigma-Aldrich®
(3-(Dimethylamino)propyl)trimethoxysilan
Formel:
(H₃C)₂NCH₂CH₂CH₂Si(OCH₃)₃ Molecular Weight: 207,35 g/mol |
MDL:
MFCD00053674 CAS-Nummer: 2530-86-1 EINECS: 219-786-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...