Butyltrimethoxysilan

Lieferant: SILANE (PART OF CHEMPUR)

SILNSILS03550.5EA 0 EUR
SILNSILS03550.5 SILNSILS03550.100 SILNSILS03550.25
Butyltrimethoxysilan
Butyltrimethoxysilan
Formel: C₇H₁₈O₃Si
Molecular Weight: 178,3 g/mol
CAS-Nummer: 1067-57-8
EINECS: 213-936-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR