Diphenylzinndichlorid
Lieferant: EHRENSTORFER GMBH
Synonyme:
Dichlordiphenylstannan
EHERC12921000EA
51
EUR
EHERC12921000
Diphenylzinndichlorid
Diphenylzinndichlorid
Formel:
C₁₂H₁₀Cl₂Sn Molecular Weight: 343,83 g/mol |
MDL:
MFCD00000516 CAS-Nummer: 1135-99-5 EINECS: 214-496-3 UN: 3146 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...