Diphenylzinndichlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dichlordiphenylstannan

M-Clarity™ quality level = MQ100

SIAL229202-25GEA 106 EUR
SIAL229202-25G SIAL229202-5G
Diphenylzinndichlorid, Sigma-Aldrich®
Diphenylzinndichlorid
Formel: C₁₂H₁₀Cl₂Sn
Molecular Weight: 343,83 g/mol
MDL: MFCD00000516
CAS-Nummer: 1135-99-5
EINECS: 214-496-3
UN: 3146
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR