Diphenylzinndichlorid ≥96%
Lieferant: Strem Chemicals, Inc.
|
Achtung
|
Synonyme:
Dichlordiphenylstannan
STRE93-5016-10GEA
69.4
EUR
STRE93-5016-10G
Diphenylzinndichlorid ≥96%
Diphenylzinndichlorid
Formel:
C₁₂H₁₀Cl₂Sn Molecular Weight: 343,83 g/mol Siedepunkt: 333…337 °C (Zers.) Schmelzpunkt: 41…43 °C |
MDL:
MFCD00000516 CAS-Nummer: 1135-99-5 EINECS: 214-496-3 UN: 3146 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...