Ammoniumbromid, Sigma-Aldrich®
Lieferant: Merck
SIAL467731-50GEA
104
EUR
SIAL467731-50G
Ammoniumbromid, Sigma-Aldrich®
Ammonium bromide
Formel:
H₄BrN Molecular Weight: 97,94 g/mol Schmelzpunkt: 542 °C Dichte: 2,42 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011419 CAS-Nummer: 12124-97-9 EINECS: 235-183-8 Merck Index: 13,00505 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...